2-Amino-5-bromopyrimidine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Bromopyrimidin-2-ylamine, 5-Bromopyrimidin-2-amine
M-Clarity™ quality level = MQ100
303526-1G 303526-5G
SIAL303526-1GEA 24.6 EUR
SIAL303526-1G SIAL303526-5G
2-Amino-5-bromopyrimidine, Sigma-Aldrich®
2-Amino-5-bromopyrimidine
Formula: C₄H₄BrN₃
MW: 174 g/mol
MDL Number: MFCD00012341
CAS Number: 7752-82-1