Trimethyl orthoformate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
305472-100ML
305472-1L
SIAL305472-100MLEA
88.4
EUR
SIAL305472-100ML
SIAL305472-1L
Trimethyl orthoformate, Sigma-Aldrich®
Trimethyl orthoformate
Formula:
C₄H₁₀O₃ MW: 106.12 g/mol Boiling Pt: 101 °C (1013 hPa) Melting Pt: –53 °C Density: 0.9691 g/cm³ (20 °C) Flash Pt: 13 °C |
MDL Number:
MFCD00008483 CAS Number: 149-73-5 EINECS: 205-745-7 UN: 3272 ADR: 3,II Merck Index: 12,07012 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...