Iron(II) oxalate dihydrate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Iron (II) ethanedioate dihydrate, Oxalic acid iron (II) salt dihydrate, Ferrous oxalate dihydrate, Iron(II) oxalate hydrate
M-Clarity™ quality level = MQ100
307726-25G
307726-500G
SIAL307726-25GEA
43.5
EUR
SIAL307726-25G
SIAL307726-500G
Iron(II) oxalate dihydrate, Sigma-Aldrich®
Iron (II) oxalate dihydrate
Formula:
Fe(C₂O₄)·2H₂O MW: 179.9 g/mol Melting Pt: 190 Density: 2.28 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150040 CAS Number: 6047-25-2 EINECS: 208-217-4 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...