Benzoyl isocyanate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
308188-1G 308188-5G
SIAL308188-1GEA 50.7 EUR
SIAL308188-1G SIAL308188-5G
Benzoyl isocyanate, Sigma-Aldrich®
Benzoyl isocyanate
Formula: C₈H₅NO₂
Boiling Pt: 75 °C (0.1 mmHg)
Melting Pt: 28…29 °C
Density: 1.17 g/cm³ (20 °C)
MDL Number: MFCD00002032
CAS Number: 4461-33-0
EINECS: 224-717-5