Tetrapotassium diphosphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tetrapotassium diphosphate, Potassium diphosphate, Potassium pyrophosphate, tetra-Potassium pyrophosphate, Potassium pyrophospate
M-Clarity™ quality level = MQ200
322431-2.5KG 322431-25G 322431-500G
SIAL322431-2.5KGEA 104 EUR
SIAL322431-2.5KG SIAL322431-25G SIAL322431-500G
Tetrapotassium diphosphate, Sigma-Aldrich®
tetra-Potassium diphosphate
Formula: K₄O₇P₂
MW: 330.34 g/mol
Boiling Pt: ∼1500 °C (1013 hPa)
Melting Pt: 1090
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00011393
CAS Number: 7320-34-5
EINECS: 230-785-7
Merck Index: 12,07833