3-Phenoxyaniline, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Amino-3-phenoxybenzol

338354-25G 338354-5G
SIAL338354-25GEA 136 EUR
SIAL338354-25G SIAL338354-5G
3-Phenoxyaniline, Sigma-Aldrich®
3-Phenoxyaniline
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 116…120 °C (0.3 mmHg)
Melting Pt: 37 °C
Density: 1,1584 g/cm³ (25 °C)
MDL Number: MFCD00041891
CAS Number: 3586-12-7
EINECS: 222-715-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR