2-Iodoperfluoropropane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,1,1,2,3,3,3-Heptafluoro-2-iodopropane, Heptafluoroisopropyl iodide, Heptafluoro-2-iodopropane, 2-Iodoheptafluoropropane
M-Clarity™ quality level = MQ100
342017-25G 342017-5G
SIAL342017-25GEA 75.6 EUR
SIAL342017-25G SIAL342017-5G
2-Iodoperfluoropropane, Sigma-Aldrich®
2-Iodoperfluoropropane
Formula: C₃F₇I
MW: 295.93 g/mol
Boiling Pt: 39 °C (1013 hPa)
Melting Pt: –56 °C
Density: 2.077 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042089
CAS Number: 677-69-0
EINECS: 211-643-3