3-(Trifluoromethoxy)benzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
346489-1G 346489-5G
SIAL346489-1GEA 26.1 EUR
SIAL346489-1G SIAL346489-5G
3-(Trifluoromethoxy)benzaldehyde, Sigma-Aldrich®
3-(Trifluoromethoxy)benzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 83…86 °C (24 mmHg)
Melting Pt: 450 °C
Density: 1.33 g/cm³ (20 °C)
MDL Number: MFCD00042406
CAS Number: 52771-21-8