Dibenzothiophene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Dibenzo[b,d]thiophene
M-Clarity™ quality level = MQ100
347833-1G
347833-5G
SIAL347833-1GEA
24.6
EUR
SIAL347833-1G
SIAL347833-5G
Dibenzothiophene, Sigma-Aldrich®
Dibenzothiophene
Formula:
C₁₂H₈S MW: 184.26 g/mol Boiling Pt: 332 °C (1013 hPa) Melting Pt: 95…98 °C Density: 1.1295 g/cm³ (125 °C) Flash Pt: 170 °C |
MDL Number:
MFCD00004969 CAS Number: 132-65-0 EINECS: 205-072-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...