2-Amino-1,3-propanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol
M-Clarity™ quality level = MQ200
357898-1G 357898-5G
SIAL357898-1GEA 47.9 EUR
SIAL357898-1G SIAL357898-5G
2-Amino-1,3-propanediol, Sigma-Aldrich®
2-Amino-1,3-propanediol
Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 91…95 °C
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0