6-Methoxy-o-toluidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Amino-1-methoxy-3-methylbenzene, 6-Methyl-o-anisidine, 2-Methoxy-6-methylaniline, 6-Methoxy-o-toluidine, 2-Amino-3-methylanisole, 2-Amino-3-methoxytoluene, 1-Amino-2-methoxy-6-methylbenzene

M-Clarity™ quality level = MQ100

360090-25G 360090-5G
SIAL360090-25GEA 201 EUR
SIAL360090-25G SIAL360090-5G
6-Methoxy-o-toluidine, Sigma-Aldrich®
6-Methoxy-o-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 26…29 °C
Density: 1.039 g/cm³ (25 °C)
MDL Number: MFCD00075455
CAS Number: 50868-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR