1H,1H,2H,2H-Perfluoro-1-iodooctane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 1-Iodo-3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane
M-Clarity™ quality level = MQ100
370517-100G 370517-25G 370517-5G
SIAL370517-100GEA 131 EUR
SIAL370517-100G SIAL370517-25G SIAL370517-5G
1H,1H,2H,2H-Perfluoro-1-iodooctane, Sigma-Aldrich®
1H,1H,2H,2H-Perfluoro-1-iodooctane
Formula: C₈H₄F₁₃I
MW: 474 g/mol
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: 20.5 °C
Density: 1.91 g/cm³ (25 °C)
MDL Number: MFCD00039410
CAS Number: 2043-57-4
EINECS: 218-056-1