2-Amino-4,6-dimethoxypyrimidine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4,6-Dimethoxypyrimidin-2-amine, 4,6-Dimethoxypyrimidin-2-ylamine
MQ100
375349-25G 375349-5G
SIAL375349-25GEA 63.1 EUR
SIAL375349-25G SIAL375349-5G
2-Amino-4,6-dimethoxypyrimidine, Sigma-Aldrich®
2-Amino-4,6-dimethoxypyrimidine
Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Boiling Pt: 276
Melting Pt: 97
Density: 1.429 g/cm³ (–153 °C)
MDL Number: MFCD00038832
CAS Number: 36315-01-2
EINECS: 252-969-6