N-Methylmaleimide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Methyl-1H-pyrrole-2,5-dione

M-Clarity™ quality level = MQ200

389412-25G 389412-5G
SIAL389412-25GEA 151 EUR
SIAL389412-25G SIAL389412-5G
N-Methylmaleimide, Sigma-Aldrich®
N-Methylmaleimide
Formula: C₅H₅NO₂
MW: 111.1 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: 70…75 °C
MDL Number: MFCD00005508
CAS Number: 930-88-1
EINECS: 213-226-1
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR