Methylamine 2 mol/l in methanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Aminomethane
M-Clarity™ quality level = MQ200
395048-100ML 395048-800ML
SIAL395048-100MLEA 119 EUR
SIAL395048-100ML SIAL395048-800ML
Methylamine 2 mol/l in methanol, Sigma-Aldrich®
Methylamine
Formula: CH₅N
Storage Temperature: Ambient
MDL Number: MFCD00008104
CAS Number: 74-89-5
EINECS: 200-820-0
UN: 1992
ADR: 3,II