Tris(4-methoxyphenyl)phosphine, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tris(p-methoxyphenyl)phosphine

M-Clarity™ quality level = MQ200

395102-1G 395102-5G
SIAL395102-1GEA 54.9 EUR
SIAL395102-1G SIAL395102-5G
Tris(4-methoxyphenyl)phosphine, Sigma-Aldrich®
Tris(4-methoxyphenyl)phosphine
Formula: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Boiling Pt: 446
Melting Pt: 135
MDL Number: MFCD00014896
CAS Number: 855-38-9
EINECS: 212-723-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR