1-Methylbenzimidazole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Methyl-1H-benzimidazole

399353-25G 399353-5G
SIAL399353-25GEA 118 EUR
SIAL399353-25G SIAL399353-5G
1-Methylbenzimidazole, Sigma-Aldrich®
1-Methylbenzimidazole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 59…62 °C
Density: 1,11 g/cm³ (25 °C)
MDL Number: MFCD00192275
CAS Number: 1632-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR