(±)-2-Methyltetrahydrofuran, Sigma-Aldrich®

Supplier: Merck
Danger


MQ100

414247-100ML 414247-1L 414247-2L
SIAL414247-100MLEA 91.1 EUR
SIAL414247-100ML SIAL414247-1L SIAL414247-2L
(±)-2-Methyltetrahydrofuran, Sigma-Aldrich®
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 79
Melting Pt: –136 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: –12 °C (closed cup)
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR