Triisopropyl orthoformate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,2',2''-[Methylidynetris(oxy)]trispropane

M-Clarity™ quality level = MQ200

415146-100ML 415146-25ML
SIAL415146-100MLEA 163 EUR
SIAL415146-100ML SIAL415146-25ML
Triisopropyl orthoformate, Sigma-Aldrich®
Triisopropyl orthoformate
Formula: C₁₀H₂₂O₃
MW: 190.28 g/mol
Boiling Pt: 64 °C (1013 hPa)
Density: 0.8821 g/cm³ (20 °C)
MDL Number: MFCD00060628
CAS Number: 4447-60-3
EINECS: 224-688-9
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR