Copper(II) chloride dihydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL459097-5GEA 61.3 EUR
SIAL459097-5G
Copper(II) chloride dihydrate, Sigma-Aldrich®
Copper(II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170,48 g/mol
Melting Pt: 100 °C
Density: 2,54 g/cm³ (20 °C)
MDL Number: MFCD00149674
UN: 2802
ADR: 8,III
Merck Index: 13,02660