5-Hexenoic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-Hexenoicacid

M-Clarity™ quality level = MQ100

464929-1G 464929-5G
SIAL464929-1GEA 56.6 EUR
SIAL464929-1G SIAL464929-5G
5-Hexenoic acid, Sigma-Aldrich®
5-Hexenoic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 107 °C (17 mmHg)
Melting Pt: –37 °C
Density: 0.961 g/cm³ (20 °C)
MDL Number: MFCD00046558
CAS Number: 1577-22-6
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR