2,2'-Butyliminodiethanol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: N-Butyldiethanolamine, N,N-Diethanolbutylamine, N-N-Butyldiethanolamine, N-Butyl-2,2'-iminodiethanol

M-Clarity™ quality level = MQ100

471240-500ML 471240-2L
SIAL471240-500MLEA 55.4 EUR
SIAL471240-500ML SIAL471240-2L
2,2'-Butyliminodiethanol, Sigma-Aldrich®
2,2'-Butyliminodiethanol
Formula: H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.9681 g/cm³ (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00002856
CAS Number: 102-79-4
EINECS: 203-055-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR