3,3,3-Trifluoropropionic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,3,3-Trifluoropropanoic acid
M-Clarity™ quality level = MQ200
498203-25ML 498203-5ML
SIAL498203-25MLEA 633 EUR
SIAL498203-25ML SIAL498203-5ML
3,3,3-Trifluoropropionic acid, Sigma-Aldrich®
3,3,3-Trifluoropropionic acid
Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.441 g/cm³ (27 °C)
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8,II