Cyclopentyl isocyanate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
516589-1G
516589-5G
SIAL516589-1GEA
64.7
EUR
SIAL516589-1G
SIAL516589-5G
Cyclopentyl isocyanate, Sigma-Aldrich®
Cyclopentyl isocyanate
Formula:
C₆H₉NO MW: 111.14 g/mol |
MDL Number:
MFCD02093695 CAS Number: 4747-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...