1-Ethyl-3-methylimidazolium chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride
53096-25G 53096-5G
SIAL53096-25GEA 245 EUR
SIAL53096-25G SIAL53096-5G
1-Ethyl-3-methylimidazolium chloride, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium chloride
Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
Melting Pt: ∼80 °C
Density: ∼1.1 g/cm³ (20 °C)
Flash Pt: 186 °C
MDL Number: MFCD00074843
CAS Number: 65039-09-0