2-Amino-6-bromobenzothiazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
538507-1G
538507-25G
538507-5G
SIAL538507-1GEA
29.3
EUR
SIAL538507-1G
SIAL538507-25G
SIAL538507-5G
2-Amino-6-bromobenzothiazole, Sigma-Aldrich®
2-Amino-6-bromobenzothiazole
Formula:
C₇H₅BrN₂S MW: 229.1 g/mol |
MDL Number:
MFCD00152229 CAS Number: 15864-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...