Zinc bromide dihydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
546739-100G 546739-500G
SIAL546739-100GEA 58.8 EUR
SIAL546739-100G SIAL546739-500G
Zinc bromide dihydrate, Sigma-Aldrich®
Zinc bromide dihydrate
Formula: Br₂Zn·2H₂O
MW: 261.23 g/mol
Storage Temperature: Ambient
CAS Number: 18921-13-6
EINECS: 231-718-4
UN: 3260
ADR: 8 III