4-Bromo-2-fluorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

554235-1G 554235-5G
SIAL554235-1GEA 25 EUR
SIAL554235-1G SIAL554235-5G
4-Bromo-2-fluorobenzenesulphonyl chloride, Sigma-Aldrich®
4-Bromo-2-fluorobenzenesulphonyl chloride
Formula: C₆H₃BrClFO₂S
MW: 273.51 g/mol
Boiling Pt: 84 °C (0.5 mmHg)
Melting Pt: 63…66 °C
MDL Number: MFCD01090993
CAS Number: 216159-03-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR