3-Methoxybenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Methoxybenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

556165-1G 556165-5G
SIAL556165-1GEA 92.7 EUR
SIAL556165-1G SIAL556165-5G
3-Methoxybenzenesulphonyl chloride, Sigma-Aldrich®
3-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 86…98 °C (1 mmHg)
Density: 1.46 g/cm³ (20 °C)
MDL Number: MFCD01318155
CAS Number: 10130-74-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR