1-(2-Bromoethyl)naphthalene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

559318-1G 559318-5G
SIAL559318-1GEA 94.7 EUR
SIAL559318-1G SIAL559318-5G
1-(2-Bromoethyl)naphthalene, Sigma-Aldrich®
1-(2-Bromoethyl)naphthalene
Formula: C₁₂H₁₁Br
MW: 235.12 g/mol
MDL Number: MFCD00037737
CAS Number: 13686-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR