(Iodomethyl)trimethylsilane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

57903-25ML 57903-5ML
SIAL57903-25MLEA 379 EUR
SIAL57903-25ML SIAL57903-5ML
(Iodomethyl)trimethylsilane, Sigma-Aldrich®
(Iodomethyl)trimethylsilane
Formula: C₄H₁₁ISi
MW: 214.12 g/mol
Boiling Pt: 139 °C (1013 hPa)
Density: 1.4301 g/cm³ (20 °C)
MDL Number: MFCD00001077
CAS Number: 4206-67-1
EINECS: 224-123-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR