2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
58030-1G-F
58030-25G-F
58030-5G-F
SIAL58030-1G-FEA
213
EUR
SIAL58030-1G-F
SIAL58030-25G-F
SIAL58030-5G-F
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride, Sigma-Aldrich®
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Formula:
C₁₉H₁₃ClIN₅O₂ Melting Pt: 227…229 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011961 CAS Number: 146-68-9 EINECS: 205-676-2 |