5-Bromo-2-chloropyrimidine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
596949-1G
596949-5G
SIAL596949-1GEA
25.1
EUR
SIAL596949-1G
SIAL596949-5G
5-Bromo-2-chloropyrimidine, Sigma-Aldrich®
5-Bromo-2-chloropyrimidine
Formula:
C₄H₂BrClN₂ MW: 193.43 g/mol |
MDL Number:
MFCD00483232 CAS Number: 32779-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...