4-Bromo-o-phenylenediamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Bromo-3,4-diaminobenzene, 4-Bromobenzene-1,2-diamine, 4-Bromo-1,2-phenylenediamine, 4-Bromo-1,2-diaminobenzene, 2-Amino-4-bromophenylamine

M-Clarity™ quality level = MQ100

640441-100G 640441-25G 640441-5G
SIAL640441-100GEA 207 EUR
SIAL640441-100G SIAL640441-25G SIAL640441-5G
4-Bromo-o-phenylenediamine, Sigma-Aldrich®
4-Bromo-o-phenylenediamine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD02660622
CAS Number: 1575-37-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR