1-(4-Fluorobenzyl)piperazin, Sigma-Aldrich®

Supplier: Merck

646164-1G 646164-5G
SIAL646164-1GEA 26.1 EUR
SIAL646164-1G SIAL646164-5G
1-(4-Fluorobenzyl)piperazin, Sigma-Aldrich®
1-(4-Fluorobenzyl)piperazin
Formula: C₁₁H₁₅FN₂
MW: 194.25 g/mol
MDL Number: MFCD01075234
CAS Number: 70931-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR