1-(2,3-Difluorophenyl)-2-thiourea, Sigma-Aldrich®

Supplier: Merck

Synonyms: N-(2,3-Difluorophenyl)thiourea, 1-(2,3-Difluorophenyl)thiourea, 2,3-Difluorophenylthiourea

M-Clarity™ quality level = MQ100

654833-1G 654833-5G
SIAL654833-1GEA 81.6 EUR
SIAL654833-1G SIAL654833-5G
1-(2,3-Difluorophenyl)-2-thiourea, Sigma-Aldrich®
1-(2,3-Difluorophenyl)-2-thiourea
Formula: (C₆H₃F₂)NC(S)NH₂
MW: 188.2 g/mol
MDL Number: MFCD07368705
CAS Number: 572889-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR