2-Amino-6-(trifluoromethyl)pyridine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL660574-100MGEA 58.2 EUR
SIAL660574-100MG SIAL660574-500MG
2-Amino-6-(trifluoromethyl)pyridine, Sigma-Aldrich®
2-Amino-6-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162,11 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 78…80 °C
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR