2-Bromo-5-(trifluoromethyl)pyridine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

661120-1G 661120-5G
SIAL661120-1GEA 69.5 EUR
SIAL661120-1G SIAL661120-5G
2-Bromo-5-(trifluoromethyl)pyridine, Sigma-Aldrich®
2-Bromo-5-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
MDL Number: MFCD00153086
CAS Number: 50488-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR