4,4-Dimethylcyclohexanone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4,4-Dimethylcyclohexan-1-one

M-Clarity™ quality level = MQ100

663654-1G 663654-250MG
SIAL663654-1GEA 42.7 EUR
SIAL663654-1G SIAL663654-250MG
4,4-Dimethylcyclohexanone, Sigma-Aldrich®
4,4-Dimethylcyclohexanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
MDL Number: MFCD00234965
CAS Number: 4255-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR