5-Methyl-1-hexyne-3-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Methyl-1-hexyn-3-ol, 5-Methylhex-1-yn-3-ol, 3-Hydroxy-5-methyl-1-hexyne
M-Clarity™ quality level = MQ100
666971-1G
666971-5G
SIAL666971-1GEA
54.9
EUR
SIAL666971-1G
SIAL666971-5G
5-Methyl-1-hexyne-3-ol, Sigma-Aldrich®
5-Methyl-1-hexyne-3-ol
Formula:
C₇H₁₂O MW: 112.17 g/mol Boiling Pt: 150…156 °C (1013 hPa) Density: 0.8618 g/cm³ (20 °C) |
MDL Number:
MFCD00041606 CAS Number: 61996-79-0 UN: 1987 ADR: 3,III |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...