2,4-Dibromothiazole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,4-dibromo-1,3-thiazole

M-Clarity™ quality level = MQ100

677914-1G 677914-5G
SIAL677914-1GEA 26.1 EUR
SIAL677914-1G SIAL677914-5G
2,4-Dibromothiazole, Sigma-Aldrich®
2,4-Dibromothiazole
Formula: C₃HBr₂NS
MW: 242.92 g/mol
MDL Number: MFCD01318994
CAS Number: 4175-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR