Allyl cyanoacetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Cyanoacetic acid Allyl Ester, Allyl Cyanoacetate, Allyl 2-cyanoacetate, Cyanoacetic acid allyl ester

M-Clarity™ quality level = MQ100

691186-1KG 691186-200KG 691186-25KG
SIAL691186-1KGEA 888 EUR
SIAL691186-1KG SIAL691186-200KG SIAL691186-25KG
Allyl cyanoacetate, Sigma-Aldrich®
Allyl cyanoacetate
Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: –40 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00013817
CAS Number: 13361-32-5
EINECS: 236-423-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR