2-Aminonicotinonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine
M-Clarity™ quality level = MQ100
697133-1G
697133-5G
SIAL697133-1GEA
37.5
EUR
SIAL697133-1G
SIAL697133-5G
2-Aminonicotinonitrile, Sigma-Aldrich®
2-Aminonicotinonitrile
Formula:
C₆H₅N₃ MW: 119.13 g/mol Boiling Pt: 268 °C (1013 hPa) Melting Pt: 134 °C |
MDL Number:
MFCD00128867 CAS Number: 24517-64-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...