1-Butyl-3-methylimidazoliumhexafluorophosphate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Butyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V), 1-(But-1-yl)-3-methyl-1H-imidazol-3-ium hexafluorophosphate
70956-250G
70956-50G
70956-5G
SIAL70956-250GEA
1030
EUR
SIAL70956-250G
SIAL70956-50G
SIAL70956-5G
1-Butyl-3-methylimidazoliumhexafluorophosphate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium hexafluorophosphate
Formula:
C₈H₁₅F₆N₂P MW: 284.18 g/mol Boiling Pt: ∼200 °C (1013 hPa) Melting Pt: ∼6 °C Density: 1.38 g/cm³ (20 °C) Flash Pt: >110 °C |
MDL Number:
MFCD03093295 CAS Number: 174501-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...