Neutral red, Sigma-Aldrich®
Supplier: Merck
Synonyms:
N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
M-Clarity™ quality level = MQ100
72210-100G
72210-25G
72210-5G
SIAL72210-100GEA
218
EUR
SIAL72210-100G
SIAL72210-25G
SIAL72210-5G
Neutral red, Sigma-Aldrich®
Neutral red
Formula:
C₁₅H₁₇ClN₄ Melting Pt: 290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012651 CAS Number: 553-24-2 EINECS: 209-035-8 Merck Index: 12,06571 |