Trimethylsilyl cyanide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethylsilylcarbonitrile
73328-250ML-F 73328-50ML-F
SIAL73328-250ML-FEA 812 EUR
SIAL73328-250ML-F SIAL73328-50ML-F
Trimethylsilyl cyanide, Sigma-Aldrich®
Trimethylsilyl cyanide
Formula: C₄H₉NSi
MW: 99.21 g/mol
Boiling Pt: 114
Melting Pt: 11
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 1
MDL Number: MFCD00001765
CAS Number: 7677-24-9
EINECS: 231-657-3
UN: 3383
ADR: 6.1 I