Phthalimide potassium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-Potassium phthalimide, Potassium 1,3-dioxoisoindolin-2-ide, Potassium phthalimide
MQ200
79790-100G 79790-500G
SIAL79790-100GEA 32.7 EUR
SIAL79790-100G SIAL79790-500G
Phthalimide potassium salt, Sigma-Aldrich®
Phthalimide potassium salt
Formula: C₈H₄KNO₂
MW: 185.22 g/mol
Melting Pt: > 300 °C
Density: 1.634 g/cm³ (20 °C)
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6