5-Bromouracil, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Bromopyrimidine-2,4(1H,3H)-dione, Bromouracil
M-Clarity™ quality level = MQ100
852473-25G
852473-5G
SIAL852473-25GEA
74.7
EUR
SIAL852473-25G
SIAL852473-5G
5-Bromouracil, Sigma-Aldrich®
5-Bromouracil
Formula:
C₄H₃BrN₂O₂ MW: 190.98 g/mol Boiling Pt: 384 °C (1013 hPa) Melting Pt: 165 °C |
MDL Number:
MFCD00006017 CAS Number: 51-20-7 EINECS: 200-084-0 Merck Index: 13,01425 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...