1-Butyl-3-methylimidazolium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride
94128-250G 94128-5G 94128-50G
SIAL94128-250GEA 444 EUR
SIAL94128-250G SIAL94128-5G SIAL94128-50G
1-Butyl-3-methylimidazolium chloride, Sigma-Aldrich®
1-Butyl-3-methylimidazolium chloride
Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼60 °C
Density: ∼1 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD03095425
CAS Number: 79917-90-1