3-Acetylindole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Indolyl methyl ketone, 1-(3-Indolyl)-1-ethanone, 1-(1H-Indol-3-yl)ethanone, Indol-3-yl methyl ketone

M-Clarity™ quality level = MQ100

A16807-25G A16807-5G
SIALA16807-25GEA 338 EUR
SIALA16807-25G SIALA16807-5G
3-Acetylindole, Sigma-Aldrich®
3-Acetylindole
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 144…145 °C (10 mmHg)
Melting Pt: 186…189 °C
MDL Number: MFCD00005626
CAS Number: 703-80-0
EINECS: 211-875-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR